missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid 47-51%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™

Catalog No. p-320640
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2 L
Packaging:
FEP Bottle
PFA Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A463250 250 mL PFA Bottle
A463500 500 mL PFA Bottle
A4631 1 L PFA Bottle
A4632 2 L FEP Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A463250 Supplier Seastar Chemicals Inc Supplier No. S020502TFIF01
Only null left

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228
IUPAC Name hydrogen fluoride
SMILES F

Specifications

pH 2.0
Color Colorless
Melting Point -35°C
Boiling Point 105°C
Packaging PFA Bottle
Certificate of Analysis Certificate of Lot included with each bottle.
Quantity 250 mL
Physical Form Liquid
UN Number 1790
Merck Index 15, 4827
Formula Weight 20.01
Grade OPTIMA
Chemical Name or Material Hydrofluoric Acid
Show More Show Less
DOTInformation DOT Class 8, : Corrosive