missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric acid, ACS reagent, 48-51% solution in water, Thermo Scientific Chemicals

Catalog No. AC423800025
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423800025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC423800025 Supplier Thermo Scientific Chemicals Supplier No. 423800025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7664-39-3 , 7732-18-5
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228
IUPAC Name hydrogen fluoride
SMILES F

Specifications

Melting Point -35.0°C
Boiling Point 105.0°C
Density 1.1500g/mL
Heavy Metals (as Pb) 0.5ppm max.
Packaging Plastic bottle
Residue after Ignition 0.0005% max.
Specific Gravity 1.15
Quantity 2.5 kg
Linear Formula HF
Solubility Information Solubility in water: soluble.
Formula Weight 20
Grade ACS Reagent
Chemical Name or Material Hydrofluoric acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement Fatal in contact with skin. Causes severe skin burns and eye damage. Fatal if swallowed. Fatal if inhaled. May be corrosive to metals.
Precautionary Statement GHS P Statement Do not breathe dust/fume/gas/mist/vapors/spray. Do not get in eyes, on skin, or on clothing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only