missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, 70%, Technical, Spectrum™ Chemical

Catalog No. 18603248
Change view
Click to view available options
Quantity:
1 gal.
4 x 1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18603248 1 gal.
18603249 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18603248 Supplier Spectrum Chemical Mfg Cor Supplier No. H10551GL
Get Up to $500 in Chemicals with Purchase
View all promotions

H1055, 7664-39-3, HF

Spectrum™ Chemical Hydrofluoric Acid, 70 Percent, Technical is a highly corrosive solution of hydrogen fluoride in water is capable of dissolving a wide range of materials including oxides and glass. Used in a variety of fields including oil refining and production of fluorides it can also be used to dissolve rock samples before analysis.

Chemical Identifiers

CAS 7664-39-3 , 7732-18-5
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
IUPAC Name hydrogen fluoride
SMILES F

Specifications

CAS Max % 60 to 80%, 20 to 40%
Quantity 1 gal.
UN Number UN1790
Percent Purity 70%
Grade Technical
Chemical Name or Material Hydrofluoric Acid