missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, 5% v/v, Reagents

Catalog No. 01335007
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335007 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335007 Supplier Labchem Inc Supplier No. CH1300004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrofluoric Acid, 5% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7664-39-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen fluoride, Fluorhydric Acid
IUPAC Name water
SMILES O

Specifications

pH <2
Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0098
Quantity 4 L
Physical Form Liquid
Percent Purity 5%
Grade Reagent
Chemical Name or Material Hydrofluoric Acid, 5% v/v