missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, 10% (v/v), Ricca Chemical

Catalog No. R38198001A
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R38198001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R38198001A Supplier Ricca Chemical Company Supplier No. R38198001A

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7664-39-3
Molecular Formula HF
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
IUPAC Name hydrogen fluoride
SMILES F

Specifications

Appearance of Solution Colorless Liquid
CAS Max % 5.43, 94.57
Packaging Natural Poly Bottle
Quantity 1 L
Percent Purity 10% V/V
Grade Laboratory
Chemical Name or Material Hydrofluoric Acid
ShelfLife 24 months