missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, 1% (w/v), Ricca Chemical

Catalog No. R38161001A
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R38161001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R38161001A Supplier Ricca Chemical Company Supplier No. R38161001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5 , 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228
IUPAC Name hydrogen fluoride
SMILES F

Specifications

Color Colorless
CAS Min % 96.0, 1.92
CAS Max % 100.0, 2.0
Packaging Natural Poly Bottle
Quantity 1 L
Physical Form Liquid
Percent Purity 1% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1% (w/v) Aqueous Solution
Chemical Name or Material Hydrofluoric Acid
ShelfLife 24 months