missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone Acetate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014336
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014336 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014336 Supplier Merck Emd Millipore Supplier No. PHR16061G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Hydrocortisone Acetate belongs to the class of corticosteroids, exhibiting antipruritic, anti-inflammatory and vasoconstrictive properties. It is used in the treatment of various conditions such as allergic and breathing disorders or skin conditions. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 50-03-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C23H32O6
MDL Number MFCD00037714
Melting Point 223°C
Synonym Hydrocortisone 21-acetate; 11 beta,17 alpha,21-Trihydroxy-4-pregnene-3,20-dione 21-acetate; 17-alpha-Hydroxycorticosterone acetate; 17-Hydroxycorticosterone 21-acetate; 21-Acetoxy-4-pregnene-11 beta,17 alpha-diol-3,20-dione; 4-Pregnene-11 beta,17 alpha,21-triol-3,20-dione 21-acetate
Type Certified Reference Material
Quantity 1 g
Formula Weight 404.50
Grade Certified Reference Material