missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone 21-hemisuccinate, MilliporeSigma™ Supelco™

Catalog No. 111013565
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013565 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013565 Supplier Merck Emd Millipore Supplier No. H28821G
Pharmaceutical (small molecule)

Specifications

Packaging Poly Bottle
CAS 2203-97-6
Physical Form Neat
Linear Formula C25H34O8
MDL Number MFCD00046256
Synonym 11 beta,17 alpha,21-Trihydroxy-4-pregnene-3,20-dione 21-hemisuccinate; Cortisol 21-hemisuccinate
Type Analytical Standard
Quantity 1 g
Formula Weight 462.53
Grade Analytical Standard