missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrocortisone 17-Butyrate 98.0+%, TCI America™
$77.00 - $240.64
Chemical Identifiers
| CAS | 13609-67-1 |
|---|---|
| Molecular Formula | C25H36O6 |
| Molecular Weight (g/mol) | 432.557 |
| MDL Number | MFCD00083364 |
| InChI Key | BMCQMVFGOVHVNG-TUFAYURCSA-N |
| Synonym | 11beta,17,21-Trihydroxypregn-4-ene-3,20-dione 17-Butyrate, Cortisol 17-Butyrate |
| PubChem CID | 26133 |
| ChEBI | CHEBI:31674 |
| IUPAC Name | [(8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] butanoate |
| SMILES | CCCC(=O)OC1(CCC2C1(CC(C3C2CCC4=CC(=O)CCC34C)O)C)C(=O)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H1506200MG
|
TCI America
H1506200MG |
200 mg |
Each for $77.00
|
|
|||||
|
H15061G
|
TCI America
H15061G |
1 g |
Each for $240.64
|
|
|||||
Chemical Identifiers
| 13609-67-1 | |
| 432.557 | |
| BMCQMVFGOVHVNG-TUFAYURCSA-N | |
| 26133 | |
| [(8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] butanoate |
| C25H36O6 | |
| MFCD00083364 | |
| 11beta,17,21-Trihydroxypregn-4-ene-3,20-dione 17-Butyrate, Cortisol 17-Butyrate | |
| CHEBI:31674 | |
| CCCC(=O)OC1(CCC2C1(CC(C3C2CCC4=CC(=O)CCC34C)O)C)C(=O)CO |
Specifications
| 13609-67-1 | |
| White | |
| MFCD00083364 | |
| 11beta,17,21-Trihydroxypregn-4-ene-3,20-dione 17-Butyrate, Cortisol 17-Butyrate | |
| CCCC(=O)OC1(CCC2C1(CC(C3C2CCC4=CC(=O)CCC34C)O)C)C(=O)CO | |
| 432.557 | |
| CHEBI:31674 | |
| ≥98.0% (HPLC) | |
| Hydrocortisone 17-Butyrate |
| 212°C | |
| C25H36O6 | |
| 200 mg | |
| BMCQMVFGOVHVNG-TUFAYURCSA-N | |
| [(8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] butanoate | |
| 26133 | |
| 432.56 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : GM9002000
TSCA : No