missing translation for 'onlineSavingsMsg'
Learn More

Hydrochlorothiazide 97.0+%, TCI America™

Catalog No. p-7130954
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12745G 5 g
H127425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12745G Supplier TCI America Supplier No. H12745G
Only null left

Chemical Identifiers

CAS 58-93-5
Molecular Formula C7H8ClN3O4S2
Molecular Weight (g/mol) 297.73
MDL Number MFCD00051765
InChI Key JZUFKLXOESDKRF-UHFFFAOYSA-N
Synonym hydrochlorothiazide, hypothiazide, esidrix, hctz, oretic, dichlotiazid, hidrotiazida, hydrochlorothiazid, hydrochlorthiazide, hydrodiuretic
PubChem CID 3639
ChEBI CHEBI:5778
IUPAC Name 6-chloro-1,1-dioxo-3,4-dihydro-2H-1λ⁶,2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)C1=CC2=C(NCNS2(=O)=O)C=C1Cl

Specifications

Melting Point 269°C
Color White-Yellow
Quantity 5 g
Formula Weight 297.73
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Hydrochlorothiazide
RTECSNumber DK9100000
TSCA Yes