missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, Ultratrace, 32-35%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor H11351LTTF
Specifications
| 7647-01-0,7732-18-5 | |
| 1 L | |
| MFCD00011324 MFCD00792839 | |
| Cl | |
| 36.46 | |
| High Purity Solvent |
| PTFE Bottle | |
| ClH | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 32 to 35% |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| Cl |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrogen chloride |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.