missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, TraceSELECT™ Ultra, Honeywell Fluka™

Catalog No. 6002970
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002970 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002970 Supplier Honeywell Inc Supplier No. 962081L
Only null left

TraceSELECT™ Ultra, for ultratrace analysis, 32-35%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Yellow
Concentration 32 to 35% (T)
Density 1.2g/mL at 25°C, 1.15g/mL at 20°C
Vapor Pressure 3.23 psi (21.1°C), 7.93 psi (37.7°C)
Quantity 1 L
UN Number UN1789
Beilstein 1098214
Formula Weight 36.46g/mol
Grade TraceSELECT™ Ultra
Chemical Name or Material Hydrochloric acid
Hazard Statement P261-P280-P305 + P351 + P338-P310
RTECSNumber MW4025000