missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, TraceSELECT™, 30.0 to 32.0 %, Honeywell Fluka™

Catalog No. p-7120474
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044608 100 mL Plastic Bottle
60044609 1 L Plastic Bottle
60044610 2.5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60044608 Supplier Honeywell Inc Supplier No. 08256100ML
Only null left

TraceSELECT™, for trace analysis,≥30%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless
Melting Point -35°C
Boiling Point 42°C
Density 1.15g/mL at 20°C, 1.2g/mL at 25°C
Packaging Plastic Bottle
Vapor Pressure 3.23psi (21.1°C), 7.93psi (37.7°C)
Quantity 100 mL
Physical Form Liquid
UN Number UN1789
Beilstein 1098214
Solubility Information Soluble in water
Formula Weight 36.46g/mol
Percent Purity 30.0 to 32.0 %
Grade TraceSELECT
Chemical Name or Material Hydrochloric Acid
Show More Show Less
Hazard Statement P261-P280-P305 + P351 + P338-P310
Precautionary Statement H290-H314-H335
DOTInformation 8/PGII
EINECSNumber 231-595-7
RTECSNumber MW4025000