missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Solution, NF, EP, 0.5 N, Spectrum™ Chemical
$1014.20 - $14292.09
Chemical Identifiers
| CAS | 7647-01-0 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| IUPAC Name | hydrogen chloride |
| SMILES | Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
18-614-908
|
Spectrum Chemical Mfg Cor
H98024LT |
4 L |
Each for $1,014.20
|
|
|||||
|
18-614-909
|
Spectrum Chemical Mfg Cor
H98024LTCS4 |
4 x 4 L |
Each for $2,889.31
|
|
|||||
|
18-614-910
|
Spectrum Chemical Mfg Cor
H980220LT |
20 L |
Each for $2,343.54
|
|
|||||
|
18-614-911
|
Spectrum Chemical Mfg Cor
H9802200LT |
200 L |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||