missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid solution, 0.05M, Honeywell Fluka™

Catalog No. 60046560
Change view
Click to view available options
Quantity:
6 x 1 L
Catalog No. Quantity
60046560 6 x 1 L
1 options

Catalog No. 60046560

Supplier: Honeywell Inc 353206X1L

Only null left

Volumetric, 0.05 M HCl (0.05N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Vapor Pressure 190 mmHg (25°C)
Quantity 6 x 1 L
UN Number UN1789
IUPAC Name chlorane
Formula Weight 36.46
Chemical Name or Material Hydrochloric acid solution, volumetric, 0.05 M HCl (0.05N)
DOTInformation 8 / PGIII