missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution, 0.02N (N/50) (Certified), Fisher Chemical

Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

Supplier:  Thermo Fisher Scientific FLSA601

Catalog No. SA60-1

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

0.10
7647-01-0,7732-18-5
Pass Test
LOT 84L Potassium acid phthalate
ClH
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Certified
Hydrochloric Acid Solution, 0.02N (N/50)
0.0198 to 0.0202N
Poly Bottle
1 L
Liquid
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0

Signal Word: CAUTION!

SDS Documents

Documents

Promotions

Promotions