missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, puriss. p.a., Honeywell Fluka™
Puriss. p.a.,≥32% (T)
$111.53 - $185.80
Chemical Identifiers
| CAS | 7647-01-0 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
| PubChem CID | 313 |
| ChEBI | CHEBI:17883 |
| SMILES | Cl |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| Cl |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 |
Specifications
| Colorless | |
| 1.16g/mL at 20°C, 1.2g/mL at 25°C | |
| 1 L | |
| MFCD00011324 MFCD00792839 | |
| 1098214 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| chlorane | |
| 313 | |
| 36.46g/mol | |
| puriss. p.a. |
| 7647-01-0 | |
| 3.23 psi (21.1°C), 7.93 psi (37.7°C) | |
| ClH | |
| UN1789 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| ≥32% (T) | |
| Hydrochloric acid |
Safety and Handling
P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 + P310
RTECSNumber : MW4025000