missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid OmniTrace Ultra, MilliporeSigma™
Specifications
Specifications
| pH | <1 (20°C) |
| Packaging | PFA Bottle |
| Quantity | 1 L |
| Physical Form | Liquid |
| Solubility Information | Soluble in water |
| Formula Weight | 36.46 |
| Grade | Not Available |
| Chemical Name or Material | Hydrochloric Acid, OmniTrace Ultra™ |
Safety and Handling
| Recommended Storage | Room Temperature |