Learn More
Hydrochloric Acid, in Isopropanol, Certified, 0.1000N ±0.0010N (0.1M), LabChem™
$94.17 - $240.83
Chemical Identifiers
| CAS | 7647-01-0 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
| PubChem CID | 313 |
| ChEBI | CHEBI:17883 |
| IUPAC Name | hydrogen chloride |
| SMILES | Cl |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |
Specifications
| Colorless | |
| 7647-01-0,67-63-0 | |
| 0.1000N ±0.0010N (0.1M) | |
| 99.64,0.36 | |
| Poly Bottle | |
| Traceable to NIST | |
| Hydrogen chloride; Carbon monoxide; Carbon dioxide; May release flammable gases | |
| ClH | |
| HCl | |
| UN1993 | |
| Soluble in water | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| Certified |
| -88°C | |
| 82°C | |
| 785kg/m3 | |
| Passes Test | |
| 785kg/m3 | |
| 12°C | |
| 1 L | |
| Liquid | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 36.46 | |
| Hydrochloric Acid, in Isopropanol |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature