missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, For determination of mercury, Reag. ISO, Reag. Ph. Eur., fuming (Hg ≤0.0000005%), ≥37%, Honeywell Fluka™
for determination of mercury, Reag. ISO, Reag. Ph. Eur., fuming (Hg ≤0.0000005%),≥37%
$154.70 - $532.60
Chemical Identifiers
| CAS | 7647-01-0 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
| PubChem CID | 313 |
| ChEBI | CHEBI:17883 |
| SMILES | Cl |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| Cl |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 |
Specifications
| Yellow | |
| ≥37% | |
| Poly bottle | |
| 3.23psi (21.1°C), 7.93psi (37.7°C) | |
| ClH | |
| 1789 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| chlorane | |
| 313 | |
| 36.46g/mol |
| 7647-01-0 | |
| 1.2g/mL at 25°C | |
| ≤0.0005% (as SO4) | |
| 2.5 L | |
| MFCD00011324 MFCD00792839 | |
| 1098214 | |
| Soluble in water | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| Hydrochloric Acid, Reag. Ph. Eur. reag. ISO |
Safety and Handling
P261-P280-P305 + P351 + P338-P310
H290-H314-H335
EINECSNumber : 231-595-7
RTECSNumber : MW4025000