missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid,OmniPure™ MilliporeSigma™
Specifications
Specifications
| Melting Point | -74°C |
| Boiling Point | 110°C |
| Packaging | HDPE Bottle |
| Assay Percent Range | 34 to 37% |
| Quantity | 2.5 L |
| Linear Formula | HCl in water |
| Formula Weight | 36.46 |
| Percent Purity | 35% |
| Grade | Not Available |
| Chemical Name or Material | Hydrochloric Acid, OmniPure™ |
Safety and Handling
| Recommended Storage | Room Temperature |