missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, Certified ACS Plus, 36.5 to 38.0%, Fisher Chemical™

Catalog No. p-4339626
Change view
Click to view available options
Quantity:
500 mL
2.5 L
19 L
6 x 2.5 L
500 lb.
Packaging:
Glass Bottle
N/A
Poly Drum
Safe-Cote™ Glass
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A144500 500 mL Glass Bottle
A144S500 500 mL Safe-Cote™ Glass
A144212 2.5 L Glass Bottle
A144C212 6 x 2.5 L Glass Bottle
A144S212 6 x 2.5 L Safe-Cote™ Glass
A144P19 19 L N/A
A144500LB 500 lb. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. A144500 Supplier Thermo Fisher Scientific Formerly Fairlawn Supplier No. FLA144500
Only null left
Edge

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH <1
Color Colorless
Melting Point -35°C
Boiling Point 57°C
Density 1.18g/cm³
Heavy Metals (as Pb) 0.1ppm max.
Identification Pass Test
Packaging Glass Bottle
Residue after Ignition 4ppm max.
Specific Gravity 1.18
Vapor Pressure 125mbar at 20°C
Quantity 500 mL
Physical Form Liquid
Solubility Information Soluble in water
Percent Purity 36.5 to 38.0% (w/w)
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:3
Flammability:0
Instability:1