missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, Certified, 5.00N ±0.05N (5.0M), LabChem™

Supplier:  LabChem LC153603

 View more versions of this product

Catalog No. LC153603


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hydrochloric Acid
7647-01-0,7732-18-5
83.04,16.96
HDPE Bottle
Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
ClH
HCl
UN1789
Soluble in water
Cl
36.46
CHEBI:17883
Certified
Colorless
5.00N ±0.05N (5.0M)
Passes Test
Traceable to NIST
2.5 L
Liquid
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
36.46

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions