Learn More
Hydrochloric Acid, Certified, 10.0% (v/v) ±0.5%, LabChem™
Supplier: LabChem LC150706
Specifications
| Hydrochloric Acid | |
| 7647-01-0,7732-18-5 | |
| Passes Test | |
| Traceable to NIST | |
| 10 L | |
| Liquid | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 36.46 | |
| Certified |
| Colorless | |
| 95.66,4.34 | |
| Cubitainer™ | |
| Hydrogen chloride; Thermal decomposition generates: Corrosive vapors | |
| ClH | |
| HCl | |
| UN1789 | |
| Soluble in water | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| 10.0% ±0.5% (v/v) |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature