missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, Certified, 0.500N ±0.005N (0.5M), LabChem™
Supplier: LabChem LC152802
Specifications
| Hydrochloric Acid, 0.5N (0.5M) | |
| 7647-01-0,7732-18-5 | |
| 98.19,1.81 | |
| Poly Bottle | |
| Hydrogen chloride; Thermal decomposition generates: Corrosive vapors | |
| ClH | |
| HCl | |
| UN1789 | |
| Soluble in water | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| Certified |
| Colorless | |
| 0.500N ± 0.005N | |
| Passes Test | |
| Traceable to NIST | |
| 1 L | |
| Liquid | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 36.46 |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/docto
Danger
Recommended Storage : Room Temperature