missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, Certified, 0.1000N ±0.0005N (0.1M), LabChem™
Supplier: LabChem LC152204
Specifications
| Hydrochloric Acid, 0.1N (0.1M) | |
| 7647-01-0,7732-18-5 | |
| 1g/mL | |
| Passes Test | |
| 1g/mL | |
| Hydrogen chloride | |
| ClH | |
| HCl | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 36.46 |
| Colorless | |
| 0.1000N ± 0.0005N | |
| 99.64,0.36 | |
| Poly Bottle | |
| Traceable to NIST | |
| 4 L | |
| Liquid | |
| MFCD00011324 MFCD00792839 | |
| Soluble in water | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| Certified |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/at
Warning
Recommended Storage : Room Temperature