missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, c(HCl)=0.1 mol/L (0.1N) Titripur™, MilliporeSigma™
Specifications
Specifications
| pH | 1.2 (H2O, 20 °C) |
| Density | 1.00 g/cm3 (20 °C) |
| Packaging | Plastic Bottle |
| Traceability to NIST | NIST SRM |
| Quantity | 1 L |
| Physical Form | Liquid |
| UN Number | 1789 |
| Grade | Not Available |
| Chemical Name or Material | Hydrochloric Acid |
Safety and Handling
| Recommended Storage | Store at 15° to 25°C |