Learn More
Hydrochloric acid, for biochemistry, approx. 37% solution in water
CAS: 7647-01-0 | ClH | 36.46 g/mol
$56.00 - $56.00
Chemical Identifiers
| CAS | 7647-01-0 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| IUPAC Name | hydrogen chloride |
| SMILES | Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC463961000
|
Thermo Scientific Chemicals
463961000 |
100 mL |
Each for $56.00
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| Cl |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrogen chloride |
Specifications
| -35.0°C | |
| 57.0°C | |
| 1.18 | |
| 1.9 mPa.s (15°C) | |
| HCl | |
| Solubility aq. soln.: miscible: 823 g/L (0°C); 561 g/L (60°C). Other solubilities: soluble in alcohol, benzene and ether, insoluble in hydrocarbons | |
| Cl | |
| 36.46 | |
| Biochemistry |
| 7647-01-0 | |
| 1.1800 g/mL | |
| 100 mL | |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 36.46 | |
| Hydrochloric acid |
Safety and Handling
GHS H Statement May be corrosive to metals.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement Keep only in original packaging Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water or shower IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 231-595-7
RUO – Research Use Only