missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, for biochemistry, approx. 37% solution in water

Catalog No. AC463965000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463965000 500 mL
AC463961000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463965000 Supplier Thermo Scientific Chemicals Supplier No. 463965000
Only null left

CAS: 7647-01-0 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Melting Point -35.0°C
Boiling Point 57.0°C
Density 1.1800 g/mL
Specific Gravity 1.18
Quantity 500 mL
Viscosity 1.9 mPa.s (15°C)
Linear Formula HCl
Solubility Information Solubility aq. soln.: miscible: 823 g/L (0°C); 561 g/L (60°C). Other solubilities: soluble in alcohol, benzene and ether, insoluble in hydrocarbons
Formula Weight 36.46
Grade Biochemistry
Chemical Name or Material Hydrochloric acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May be corrosive to metals.

Causes severe skin burns and eye damage.

May cause respiratory irritation.

Precautionary Statement GHS P Statement Keep only in original packaging Wear protective gloves/protective clothing/eye protection/face protection.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

EINECSNumber 231-595-7

RUO – Research Use Only