missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, ACS Reagent, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Density | 1.2g/mL at 25°C |
| Residue after Ignition | ≤5ppm |
| Vapor Pressure | 7.93psi (37.7°C), 3.23psi (21.1°C) |
| Quantity | 6 x 500 mL |
| UN Number | UN1789 |
| Formula Weight | 36.46 |
| Percent Purity | 37% |
| Grade | ACS Reagent |
| Chemical Name or Material | Hydrochloric Acid |
Safety and Handling
| Hazard Statement | P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 + P310 |
| Precautionary Statement | H290-H314-H335 |
| DOTInformation | 8 / PGII |
| EINECSNumber | 231-595-7 |
| RTECSNumber | MW4025000 |