missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid ACS, MilliporeSigma™

Catalog No. p-3176275
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Poly Coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MHX06034 500 mL Glass Bottle
MHX0603P5 500 mL Poly Coated Glass Bottle
MHX06033 2.5 L Glass Bottle
23749902 2.5 L Poly Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MHX06034 Supplier Technidata Supplier No. HX06034
Only null left

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Melting Point -74°C
Boiling Point 110°C
Packaging Glass Bottle
Quantity 500 mL
Linear Formula HCl in Water
Formula Weight 36.46
Grade ACS
Chemical Name or Material Hydrochloric Acid