missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid ACS, MilliporeSigma™
Specifications
Specifications
| Packaging | Poly Coated Glass Bottle |
| Quantity | 2.5 L |
| Linear Formula | HCl in water |
| Formula Weight | 36.46 |
| Grade | ACS |
| Chemical Name or Material | Hydrochloric Acid |