missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 6.0 Normal, Reagents
Hydrochloric Acid, 6.0 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
$95.63 - $1101.97
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | Hydrogen chloride solution |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-336-588
|
Labchem Inc
C710115010F |
10 L |
Each for $681.77
|
|
|||||
|
01-336-587
|
Labchem Inc
C71011501A |
1 L |
Each for $114.83
|
|
|||||
|
01-336-586
|
Labchem Inc
C710115020E7 |
20 L |
Each for $1,101.97
|
|
|||||
|
01-336-585
|
Labchem Inc
C71011504A |
4 L |
Each for $370.33
|
|
|||||
|
01-336-584
|
Labchem Inc
C7101150500A |
500 mL |
Each for $95.63
|
|
|||||
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Hydrogen chloride solution | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| <1 | |
| 7732-18-5,7647-01-0 | |
| Cubitainer | |
| 10 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent |
| Colorless to Green/Yellow | |
| 108°C | |
| 1.0993 | |
| H2O | |
| Hydrogen chloride solution | |
| O | |
| 18.02 | |
| Hydrochloric Acid, 6.0 Normal |