missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 50% (v/v), Ricca Chemical
$43.28 - $661.25
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
| PubChem CID | 313 |
| ChEBI | CHEBI:17883 |
| IUPAC Name | hydrogen chloride |
| SMILES | Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R3580000100
|
Ricca Chemical Company
R3580000100A |
100 mL | Natural Poly Bottle |
Each for $43.28
|
|
||||
|
3580-16
|
Ricca Chemical Company
358016 |
500 mL | Natural Poly Bottle |
Each for $124.81
|
|
||||
|
3580-32
|
Ricca Chemical Company
358032 |
1 L | Natural Poly Bottle |
Each for $166.00
|
|
||||
|
35805PT
|
Ricca Chemical Company
35805PT |
2.5 L | Safety-coated Glass Bottle |
Each for $234.44
|
|
||||
|
35802-5
|
Ricca Chemical Company
358025 |
10 L | Cubitainer |
Each for $661.25
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |
Specifications
| Colorless | |
| Slightly pungent to pungent | |
| Aqueous Solution | |
| 100 mL | |
| Liquid | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 50% V/V | |
| Hydrochloric Acid, 50% (v/v) |
| 7732-18-5,7647-01-0 | |
| Natural Poly Bottle | |
| 50.0 ± 0.4% (v/v) | |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| Solubility in Water: Infinite | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| Laboratory |