missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 5.00 N, Ricca Chemical

$110.34 - $1015.56

Chemical Identifiers

CAS 7732-18-5
Molecular Formula ClH
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl
View More Specs

Products 7
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
R3740000500
SDS View Documents
Ricca Chemical Company
R3740000500A
500 mL Natural Poly Bottle
Each for $115.00
Only null left
 
374032G
SDS View Documents
Ricca Chemical Company
374032G
1 L Amber Glass Bottle
Each for $110.56
Only null left
 
3740-32
SDS View Documents
Ricca Chemical Company
374032
1 L Natural Poly Bottle
Each for $110.34
Only null left
 
37405PT
SDS View Documents
Ricca Chemical Company
37405PT
2.5 L Safety-coated Glass Bottle
Each for $221.47
Only null left
 
R37400004F
View Documents
Ricca Chemical Company
R37400004F
4 L Cubitainer
Each for $333.91
Only null left
 
3740-1
SDS View Documents
Ricca Chemical Company
37401
4 L Natural Poly Bottle
Each for $315.34
Only null left
 
3740-5
SDS View Documents
Ricca Chemical Company
37405
20 L Cubitainer
Each for $1,015.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
MFCD00011324 MFCD00792839
313
hydrogen chloride
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
CHEBI:17883
Cl

Specifications

Colorless to Green-Yellow
7732-18-5,7647-01-0
4.995 to 5.005 N
Natural Poly Bottle
ClH
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Laboratory
Normal at 20°C
Colorless to slightly Greenish-yellow Liquid
66.0,32.21
68.0,33.53
500 mL
Liquid
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
Hydrochloric Acid
Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 months

SDS
missing translation for 'documents'

missing translation for 'documents'