missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 4% (v/v), Ricca Chemical
Supplier: Ricca Chemical Company 356745
Specifications
| Hydrochloric Acid | |
| 94.0,4.42 | |
| Cubitainer | |
| ClH | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| [H+].[Cl-] | |
| 36.46 | |
| CHEBI:17883 |
| 7647-01-0,7732-18-5 | |
| 97.0,4.6 | |
| 20 L | |
| MFCD00011324 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 4% (v/v) Aqueous Solution |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| [H+].[Cl-] |
Safety and Handling
ShelfLife : 24 months