missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, meets analytical spec. of BP, NF, Ph. Eur., fuming, 36.5-38%, Honeywell Fluka™
Meets analytical specification of BP, NF, Ph. Eur., fuming, 36.5-38%
Supplier: Honeywell Inc 071026X1L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Hydrochloric Acid | |
| Meets analytical specification of Ph. Eur.BP, NF | |
| 1.2g/mL at 25°C | |
| ≤0.002% (as SO4) | |
| 6 x 1 L | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| 36.5 to 38% |
| Yellow | |
| 7647-01-0 | |
| Plastic Bottle | |
| 3.23psi (21.1°C), 7.93psi (37.7°C) | |
| ClH | |
| UN1789 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 36.46 |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| Cl |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 |
Safety and Handling
P261-P280-P305 + P351 + P338-P310
H290-H314-H335
DOTInformation : 8 / PGII
EINECSNumber : 231-595-7
RTECSNumber : MW4025000