missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 3 N Solution, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor H1504LT
Specifications
| Hydrochloric Acid | |
| 3.00 N ± 0.03 N | |
| Amber Glass Bottle | |
| Ungraded |
| 7647-01-0,7732-18-5 | |
| 10.4%,89.6% | |
| 4 L |
Chemical Identifiers
| 7647-01-0 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.