missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 3.00 Normal, Reagents

Hydrochloric Acid, 3.00 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

$113.73 - $851.20

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name water
SMILES O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-335-095
View Documents
Labchem Inc
CH1032001A
1 L
Each for $113.73
Only null left
 
01-335-094
View Documents
Labchem Inc
CH10320020F
20 L
Each for $851.20
Only null left
 
01-335-093
View Documents
Labchem Inc
CH1032004A
4 L
Each for $242.40
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
18.02
Hydrogen chloride solution
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water

Specifications

<1
7732-18-5,7647-01-0
Poly Bottle
1 L
Liquid
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Reagent
Colorless to Green/Yellow
103°C
1.0515
H2O
Hydrogen chloride solution
O
18.02
Hydrochloric Acid, 3.00 Normal
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'