missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, 2N, TitriPUR™, MilliporeSigma™
Specifications
Specifications
| Color | Colorless |
| Density | 1.03g/cm3 (20°C) |
| Packaging | Plastic Bottle |
| Quantity | 1 L |
| Physical Form | Liquid |
| Solubility Information | Soluble (20°C) |
| Grade | Not Available |
| Chemical Name or Material | Hydrochloric Acid, Titripur™ |
Safety and Handling
| Hazard Category | SignalWord: Warning |
| Recommended Storage | Store at +15°C to +25°C |