missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 2.50 N, Ricca Chemical

Supplier:  Ricca Chemical Company 371532

 View more versions of this product

Catalog No. 3715-32


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hydrochloric Acid
7732-18-5,7647-01-0
2.50Normal
Natural Poly Bottle
ClH
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Laboratory
Colorless to Green-Yellow
79.0,19.31
82.0,20.1
1 L
Liquid
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883

Chemical Identifiers

7732-18-5
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions