missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 2.00 N, Ricca Chemical

Supplier:  Ricca Chemical Company 371032

 View more versions of this product

Catalog No. 3710-32


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hydrochloric Acid
7732-18-5,7647-01-0
2.00N
Natural Poly Bottle
ClH
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Laboratory
Colorless
16.05,82.0
16.71,85.0
1 L
Liquid
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
Cl
36.46
CHEBI:23116

Chemical Identifiers

7732-18-5
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
CHEBI:23116
Cl
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Accessories
Promotions

Promotions