missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 1.00 N, Ricca Chemical
Supplier: Ricca Chemical Company RI37005
Specifications
| Hydrochloric Acid | |
| 89.0,8.71 | |
| Cubitainer | |
| ClH | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| 1.00N |
| 7732-18-5,7647-01-0 | |
| 93.0,9.07 | |
| 20 L | |
| MFCD00011324 MFCD00792839 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| Laboratory |
Chemical Identifiers
| 7732-18-5 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |