missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.500 N (N/2), Ricca Chemical
Hydrochloric Acid Titrant
$78.72 - $347.97
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
| PubChem CID | 313 |
| ChEBI | CHEBI:17883 |
| IUPAC Name | hydrogen chloride |
| SMILES | Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R3650000500
|
Ricca Chemical Company
R3650000500A |
500 mL | Natural Poly Bottle |
Each for $94.09
|
|
||||
|
3650-32
|
Ricca Chemical Company
365032 |
1 L | Natural Poly Bottle |
Each for $78.72
|
|
||||
|
R36500004F
|
Ricca Chemical Company
R36500004F |
4 L | Cubitainer |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
3650-1
|
Ricca Chemical Company
RI36501 |
4 L | Natural Poly Bottle |
Each for $167.09
|
|
||||
|
3650-25
|
Ricca Chemical Company
365025 |
10 L | Cubitainer |
Each for $347.97
|
|
||||
|
3650-5
|
Ricca Chemical Company
36505 |
20 L | Cubitainer |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
RIC01001ST
|
Ricca Chemical Company
36505HP |
20 L | Hard Pack |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7732-18-5 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |
Specifications
| Colorless | |
| Odorless to slightly pungent | |
| Aqueous Solution | |
| 500 mL | |
| Liquid | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| Laboratory |
| 7732-18-5,7647-01-0 | |
| Natural Poly Bottle | |
| 0.5000 ± 0.0002 N at 20°C | |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| Solubility in Water: Infinite | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| Hydrochloric Acid, 0.5 Normal |
Safety and Handling
ShelfLife : 24 months
Recommended Storage : Ambient Corrosive