missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.300 N, Ricca Chemical
$94.81 - $94.81
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
| PubChem CID | 313 |
| ChEBI | CHEBI:17883 |
| IUPAC Name | hydrogen chloride |
| SMILES | Cl |
Chemical Identifiers
| 7732-18-5 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |
Specifications
| Colorless | |
| 95.0,2.8 | |
| 99.0,2.91 | |
| 1 L | |
| Liquid | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| Hydrochloric Acid |
| 7732-18-5,7647-01-0 | |
| 0.300Normal | |
| Natural Poly Bottle | |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| Laboratory |