missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.300 Normal, Reagents
Hydrochloric Acid, 0.300 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.
$155.57 - $464.70
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | Hydrogen chloride solution |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-336-590
|
Labchem Inc
C710109020F |
20 L |
Each for $464.70
|
|
|||||
|
01-336-589
|
Labchem Inc
C71010904A |
4 L |
Each for $155.57
|
|
|||||
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Hydrogen chloride solution | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| <1 | |
| 7732-18-5,7647-01-0 | |
| Cubitainer | |
| 20 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent |
| Colorless | |
| 100°C | |
| 1.0054 | |
| H2O | |
| Hydrogen chloride solution | |
| O | |
| 18.02 | |
| Hydrochloric Acid, 0.300 Normal |