missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.250 N (N/4), Ricca Chemical
Supplier: Ricca Chemical Company 362525
Specifications
| Hydrochloric Acid | |
| 96.0,2.34 | |
| Cubitainer | |
| ClH | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| [H+].[Cl-] | |
| 36.46 | |
| CHEBI:17883 |
| 7647-01-0,7647-01-0 | |
| 100.0,2.44 | |
| 10 L | |
| MFCD00011324 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| [H+].[Cl-] |