missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.100 N (N/10), Ricca Chemical
$64.16 - $317.53
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
| PubChem CID | 313 |
| ChEBI | CHEBI:17883 |
| IUPAC Name | hydrogen chloride |
| SMILES | Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
360016
|
Ricca Chemical Company
360016 |
500 mL | Natural Poly Bottle |
Each for $64.16
|
|
||||
|
3600-32
|
Ricca Chemical Company
360032 |
1 L | Natural Poly Bottle |
Each for $81.97
|
|
||||
|
36001CT
|
Ricca Chemical Company
36001CT |
4 L | Cubitainer |
Each for $264.84
|
|
||||
|
3600-1
|
Ricca Chemical Company
36001 |
4 L | Natural Poly Bottle |
Each for $179.34
|
|
||||
|
360025
|
Ricca Chemical Company
360025 |
10 L | Cubitainer |
Each for $298.41
|
|
||||
|
3600-5
|
Ricca Chemical Company
36005 |
20 L | Cubitainer |
Each for $317.53
|
|
||||
|
RIC01000ST
|
Ricca Chemical Company
36005HP |
20 L | Hard Pack |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7732-18-5 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |
Specifications
| 7732-18-5,7647-01-0 | |
| 100.0,0.99 | |
| 500 mL | |
| MFCD00011324 MFCD00792839 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| Laboratory | |
| Hydrochloric Acid |
| 97.0,0.95 | |
| Natural Poly Bottle | |
| ClH | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| 0.100N |
Safety and Handling
ShelfLife : 24 Months
Recommended Storage : General