missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.0500 N (N/20), Ricca Chemical

Hydrochloric Acid Titrant

Supplier:  Ricca Chemical Company 35975

 View more versions of this product

Catalog No. 3597-5


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hydrochloric Acid
7732-18-5,7647-01-0
0.0500Normal
Cubitainer
ClH
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Laboratory
Colorless
98.0,0.48
100.0,0.5
20 L
Liquid
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883

Chemical Identifiers

7732-18-5
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions