missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrobromic acid, for analysis, ca. 48% solution in water
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
| Color | Yellow |
| Melting Point | -11.0°C |
| Boiling Point | 126.0°C to 128.0°C |
| Density | 1.4800g/mL |
| Heavy Metals (as Pb) | 5ppm max. |
| Packaging | Glass bottle |
| Specific Gravity | 1.48 |
| Linear Formula | HBr |
| Quantity | 500 mL |
| Physical Form | Solution |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement May be corrosive to metals. Causes severe skin burns and eye damage. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. |
RUO – Research Use Only